Learn More
Eurofins Discoverx LTK, active, 10 µg. (14-958)
Supplier:  Eurofins Discoverx 14958
Encompass
N-terminal GST-tagged recombinant, human LTK amino acids 450-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9484