Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human LTK (TYK1), GST Tag Recombinant Protein

Catalog No. PR8108A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR8108A 100 μg
PV4651 10 μg
PV4652 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR8108A Supplier Invitrogen™ Supplier No. PR8108A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic

LTK (TYK1), ALK and Ros have been identified as receptor tyrosine kinases having sequence similarity to the Insulin receptor subfamily of kinases. LTK, leukocyte tyrosine kinase, is expressed in murine B-lymphocyte precursors and has also been found in forebrain neurons. ALK, anaplastic lymphoma kinase, is normally highly expressed specifically in the nervous system. A truncated form containing the catalytic domain of ALK is expressed as the result of a translocation occurring in many non-Hodgkin's lymphomas. The c-Ros gene was originally identified in mutant form as an oncogene. Ros is normally expressed in a small number of epithelial cell types and may play a role in epithelial development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29376
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 0.02% Triton X-100, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 4058
Molecular Weight (g/mol) 73.7 kDa
Name Human LTK (TYK1), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias EC 2.7.1.112; EC 2.7.10.1; leukocyte receptor tyrosine kinase; leukocyte tyrosine kinase; leukocyte tyrosine kinase receptor; Ltk; Protein tyrosine kinase 1; TYK1; tyrosine kinase-1
Common Name LTK
Gene Symbol LTK
Kinase Family ALK Kinase Family
Protein Length 450-864
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 450-864
Content And Storage Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain, Cytoplasmic
Form Liquid
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.