Learn More
Cayman Chemical (−)-L-threo-PDMP (hydrochloride), Purity: ≥98%, CAS: 161491-04-9, 10 mg
SDP
Catalog No. 502888000
Encompass_Preferred
An enhancer of ganglioside biosynthesis; increases the activity of ganglioside GM3, -GD3, and -GQ1b synthases in primary rat cortical neurons; increases the levels of glucosylceramide, lactosylceramide, and ganglioside GM3 in B16 melanoma cells; increases neurite outgrowth in primary rat neocortical explants at 15 µM; increases functional synapse formation in primary rat cortical neurons at 20 µM; improves retention of a long-term memory learned prior to forebrain ischemia in rats when administered after ischemia at 40 mg/kg per day
Catalog No. 50-288-8000 Supplier Cayman Chemical Supplier No. 1000527810 MG
An enhancer of ganglioside biosynthesis; increases the activity of ganglioside GM3, -GD3, and -GQ1b synthases in primary rat cortical neurons; increases the levels of glucosylceramide, lactosylceramide, and ganglioside GM3 in B16 melanoma cells; increases neurite outgrowth in primary rat neocortical explants at 15 µM; increases functional synapse formation in primary rat cortical neurons at 20 µM; improves retention of a long-term memory learned prior to forebrain ischemia in rats when administered after ischemia at 40 mg/kg per day