Learn More
Alkali Scientific PurePoint™ LTS Filter Tips, 200µL, for Rainin 2, Low Retention, 96 Tips/Rack, 10 Racks, 960 Tips/cs
SDP
Catalog No. NC2076644
Encompass_Preferred
PurePoint™ LTS Filter Tips fit Rainin* LTS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, and pyrogen free. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Stable new rack designed for multichannel pipettors. Great fit on Rainin* LiteTouchª pipettors. RNAse, DNAse, pyrogen free. Sterile.
Catalog No. NC2076644 Supplier Alkali Scientific Supplier No. FLT200
PurePoint™ LTS Filter Tips fit Rainin* LTS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, and pyrogen free. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Stable new rack designed for multichannel pipettors. Great fit on Rainin* LiteTouchª pipettors. RNAse, DNAse, pyrogen free. Sterile.