Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ LTS Compatible Pipet Tip

Catalog No. 01671771 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
1000 μL
Format:
Rack
Reload Deck
TipStation™
Sterility:
Non-sterile
Sterile
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format Sterility
01-671-771 200 μL Rack Sterile
01-671-780 1000 μL Rack Non-sterile
01-671-778 1000 μL Reload Deck Non-sterile
01-671-779 1000 μL Rack Sterile
01-671-772 200 μL Rack Non-sterile
01-671-770 200 μL Reload Deck Non-sterile
01-671-769 200 μL TipStation™ Non-sterile
01-671-768 200 μL TipStation™ Sterile
01-671-777 20 μL Rack Non-sterile
01-671-775 20 μL Reload Deck Non-sterile
01-671-776 20 μL Rack Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 01-671-771 Supplier Axygen™ Supplier No. LT200RS
Only null left
Add to Cart
Add to Cart

Ergonomically designed to reduce discomfort and optimized to work with Rainin LiteTouch Pipettors. The tips offer comfort you can feel with a reduction of force required to seal and eject, enabling a streamlined experience from aspiration to dispensing.

  • The Axygen LTS compatible tips are ergonomically designed to reduce discomfort and optimized to work with Rainin LiteTouch Pipettors
  • The tips offer comfort you can feel with a reduction of force required to seal and eject, enabling a streamlined experience from aspiration to dispensing.
  • Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting
  • Graduation lines for quick accuracy checks
  • Fine point design for crisp sample dispensing
  • Ultra Clear USP Class VI resin
  • All tips are certified RNase-/DNase-free, Human gDNA-, PCR-inhibition-free, and nonpyrogenic.
  • Tips are offered in a variety of packaging formats; racked, racked/filtered, tip station, reload stacks, and our more sustainable solution: Axygen HybridRack™ Pipet Tips.
  • Non-sterile product is autoclavable at 122°C for 15 minutes
  • Manufactured in the USA and meet ISO standards
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Standard
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Rainin LTS LiteTouch
Graduated No
Material Plastic
Show More Show Less

Unless otherwise specified, all products are for research use or general laboratory use only. * Not intended for use in diagnostic or therapeutic procedures. Not for use in humans. These products are not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment. Corning Life Sciences makes no claims regarding the performance of these products for clinical or diagnostic applications. Corning’s products are not specifically designed and tested for diagnostic testing. Many Corning products, though not specific for diagnostic testing, can be used in the workflow and preparation of the test at the customers discretion. Customers may use these products to support their claims. We cannot make any claims or statements that our products are approved for diagnostic testing either directly or indirectly. The customer is responsible for any testing, validation, and/or regulatory submissions that may be required to support the safety and efficacy of their intended application.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.