Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ LTS Compatible Pipette Tip, Racked

Catalog No. 01549043 Shop All Axygen Products
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
1250 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-549-043 20 μL
01-549-039 200 μL
01-549-029 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-549-043 Supplier Axygen™ Supplier No. LT20LR
Add to Cart
Add to Cart

Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting.

  • Ergonomically designed to reduce discomfort and optimized to work with Rainin LiteTouch pipettors all with the quality you have come to expect from Corning
  • The tips offer comfort you can feel with a reduction of force required to seal and eject enabling a streamlined experience from aspiration to dispensing
  • Avoid slamming the pipettor for an effective seal eliminate the struggle to eject tips and minimize sore hands and wrists after extensive pipetting
  • Axygen LTS compatible tips are offered in a variety of packaging formats racked rackedfiltered tip station reload stacks and Axygen HybridRack™
  • Now also available with MaxRecovery+™ our internally developed IP protected low retention technology featuring improved low retention performance and solvent compatibility with no intentionally added PFAS
  • Axygen LTS compatible tips are manufactured in the USA and meet ISO standards
  • All tips are certified RNase DNase-free Human gDNA-free PCR-inhibition-free and nonpyrogenic
  • Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting
  • Those SKUs designated as MaxRecovery+ are ideal for viscous aqueous solutions solvents DNA and protein-based samples ensuring superior sample recovery in the lab regardless of what you are pipetting
  • Graduation lines for quick accuracy checks
  • Fine point design for crisp sample dispensing
  • Ultra clear USP Class VI resin
  • Non-sterile product is autoclavable at 122°C for 15 minutes
  • Available with MaxRecovery+ our proprietary next generation low retention technology formulated with no intentionally added PFAS
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Red
Autoclavable Autoclavable
Format Rack
Tip Style Low Retention
Disposable Single-use
For Use With (Equipment) Rainin LTS LiteTouch
Graduated Yes
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.