Learn More
Selleck Chemical LLC LTURM34 5mg 1879887-96-3
SDP
Supplier:  Selleck Chemical LLC S84275MG
Encompass_Preferred
LTURM34 is a specific DNA-PK inhibitor, 170-fold more selective for DNA-PK activity compared to PI3K activity, with IC50 value of 0.034 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8257