Learn More
Medchemexpress LLC HY-101667 5mg Medchemexpress, LTURM34 CAS:1879887-96-3 Purity:>98%
SDP
Catalog No. 501932192
Encompass_Preferred
Medchemexpress, HY-101667 5mg LTURM34 CAS:1879887-96-3 LTURM34 is a specific DNA-PK inhibitor ( IC 50 =34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2192 Supplier Medchemexpress LLC Supplier No. HY1016675MG
Medchemexpress, HY-101667 5mg LTURM34 CAS:1879887-96-3 LTURM34 is a specific DNA-PK inhibitor ( IC 50 =34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.