Learn More
Medchemexpress LLC HY-18667 5mg Medchemexpress, LTV-1 CAS:347379-29-7 Purity:>98%
SDP
Catalog No. 502082063
Encompass_Preferred
Medchemexpress, HY-18667 5mg LTV-1 CAS:347379-29-7 LTV-1 is a potent lymphoid tyrosine phosphatase (LYP) inhibitor in T cells with an IC50 of 508 nM. LTV-1 has the potential for autoimmunity treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2063 Supplier Medchemexpress LLC Supplier No. HY186675MG
Medchemexpress, HY-18667 5mg LTV-1 CAS:347379-29-7 LTV-1 is a potent lymphoid tyrosine phosphatase (LYP) inhibitor in T cells with an IC50 of 508 nM. LTV-1 has the potential for autoimmunity treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.