Learn More
Cayman Chemical L-Tyrosine (sodium salt hydrate), Purity: ≥98%, 250 g
SDP
Catalog No. 502912185
Encompass_Preferred
A conditionally essential amino acid; a precursor in the biosynthesis of catecholamine neurotransmitters, melanins, and thyroid hormones; plasma, skeletal muscle, and erythrocyte levels are decreased in patients with chronic kidney disease
Catalog No. 50-291-2185 Supplier Cayman Chemical Supplier No. 36333250 G
A conditionally essential amino acid; a precursor in the biosynthesis of catecholamine neurotransmitters, melanins, and thyroid hormones; plasma, skeletal muscle, and erythrocyte levels are decreased in patients with chronic kidney disease