Learn More
Medchemexpress LLC HY-133910 5mg Medchemexpress, Lu AE98134 CAS:849000-18-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1339105MG
Encompass_Preferred
Medchemexpress, HY-133910 5mg Lu AE98134 CAS:849000-18-6 Lu AE98134, an activator of voltage-gated sodium channels, acts as a partly selective Nav1.1 channels positive modulator. Lu AE98134 also increases the activity of Nav1.2 and Nav1.5 channels but not of Nav1.4, Nav1.6 and Nav1.7 channels. Lu AE98134 can be used to analyze pathophysiological functions of the Nav1.1 channel in various central nervous system diseases, including cognitive restoring in schizophrenia, et al. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1351