Learn More
Medchemexpress LLC Lu AE98134 | 849000-18-6 | 98.1% | C21H23N5O3S | 10MG
SDP
Catalog No. 5000258501
Encompass_Preferred
Lu AE98134 is a small-molecule activator and positive modulator of Nav1.1 voltage-gated sodium channels, used in research to probe sodium channel function and central nervous system disease models. It shows selectivity for Nav1.1 while also increasing Nav1.2 and Nav1.5 activity, and is supplied with analytical data supporting research use.
  • Selective activator of Nav1.1 with activity at Nav1.2 and Nav1.5.
  • High purity suitable for research (≈98.1%).
  • High solubility in DMSO for in vitro preparation.
  • Recommended storage conditions provided for powder and solutions.
  • Available in small pack sizes for experimental use.
Catalog No. 50-002-58501 Supplier Medchemexpress LLC Supplier No. HY13391010MG
Lu AE98134 is a small-molecule activator and positive modulator of Nav1.1 voltage-gated sodium channels, used in research to probe sodium channel function and central nervous system disease models. It shows selectivity for Nav1.1 while also increasing Nav1.2 and Nav1.5 activity, and is supplied with analytical data supporting research use.
  • Selective activator of Nav1.1 with activity at Nav1.2 and Nav1.5.
  • High purity suitable for research (≈98.1%).
  • High solubility in DMSO for in vitro preparation.
  • Recommended storage conditions provided for powder and solutions.
  • Available in small pack sizes for experimental use.