Learn More
Medchemexpress LLC Lu AF21934 | 1445605-23-1 | 100.0% | 315.20 | C14H16Cl2N2O2 | 1 ML
SDP
Catalog No. 5000213284
Encompass_Preferred
Lu AF21934 is a selective, brain-penetrant positive allosteric modulator of the metabotropic glutamate receptor 4 (mGlu4). It potentiates mGlu4 signaling (EC50 500 nM) and is supplied for research use; available as solid and as a 10 mM solution in DMSO.
  • Selective positive allosteric modulator of mGlu4 receptor.
  • Brain-penetrant profile suitable for CNS research.
  • Potent activity with EC50 of 500 nM in mGlu4 assays.
  • Available as solids and as a 10 mM solution in DMSO.
  • High reported purity (99.99%) for pharmacological studies.
  • SMILES string available for cheminformatics and modeling.
Catalog No. 50-002-13284 Supplier Medchemexpress LLC Supplier No. HY10036610MM 1ML
Lu AF21934 is a selective, brain-penetrant positive allosteric modulator of the metabotropic glutamate receptor 4 (mGlu4). It potentiates mGlu4 signaling (EC50 500 nM) and is supplied for research use; available as solid and as a 10 mM solution in DMSO.
  • Selective positive allosteric modulator of mGlu4 receptor.
  • Brain-penetrant profile suitable for CNS research.
  • Potent activity with EC50 of 500 nM in mGlu4 assays.
  • Available as solids and as a 10 mM solution in DMSO.
  • High reported purity (99.99%) for pharmacological studies.
  • SMILES string available for cheminformatics and modeling.