Learn More
Medchemexpress LLC HY-100366 5mg Medchemexpress, Lu AF21934 CAS:1445605-23-1 Purity:>98%
SDP
Catalog No. 501931789
Encompass_Preferred
Medchemexpress, HY-100366 5mg Lu AF21934 CAS:1445605-23-1 Lu AF21934 is a selective and brain-penetrant mGlu4 receptor positive allosteric modulator with an IC 50 of 500 nM for human mGlu4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1789 Supplier Medchemexpress LLC Supplier No. HY1003665MG
Medchemexpress, HY-100366 5mg Lu AF21934 CAS:1445605-23-1 Lu AF21934 is a selective and brain-penetrant mGlu4 receptor positive allosteric modulator with an IC 50 of 500 nM for human mGlu4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.