Learn More
Med Vet International Surgilube Lubricating Jelly - Carbomer free, 3 Gram, Individual Packet, Sterile, 281020543, 1728/Case
Catalog No. 503043993
Encompass_Preferred
The Surgilube lubricating jelly has premium viscosity that maintains the integrity of the lube It's water-soluble and doesn't stain fabric The lubricant is pH neutral for natural skin balance It's biostatic and will not cause infections It's paraben-, dye- and latex-free for sensitive skin Surgilube is a universal lubricant that can be used topically and internally
Catalog No. 50-304-3993 Supplier Med Vet International Supplier No. 1009103CS
The Surgilube lubricating jelly has premium viscosity that maintains the integrity of the lube It's water-soluble and doesn't stain fabric The lubricant is pH neutral for natural skin balance It's biostatic and will not cause infections It's paraben-, dye- and latex-free for sensitive skin Surgilube is a universal lubricant that can be used topically and internally