Learn More
Med Vet International Lubricating Jelly - Carbomer free Surgilube 4.25 oz. Tube Sterile, 281020536, 12/Box
Catalog No. 503043821
Encompass_Preferred
The Surgilube lubricating jelly enhances comfort The high viscosity keeps surfaces smooth for easy insertion of instruments This sterile surgical lubricant contains chlorhexidine to prevent bacterial infections Comes in a 4.25 oz tube
Catalog No. 50-304-3821 Supplier Med Vet International Supplier No. 1050799BX
The Surgilube lubricating jelly enhances comfort The high viscosity keeps surfaces smooth for easy insertion of instruments This sterile surgical lubricant contains chlorhexidine to prevent bacterial infections Comes in a 4.25 oz tube