Learn More
Med Vet International Lubricating Jelly HR 4 oz. Tube Sterile, 201, Each
Catalog No. 503044735
Encompass_Preferred
Active ingredients include water, propylene glycol and hydroxypropyl methylcellulose HR lubricating jelly is chlorhexidine free Bacteriostatic HR lubricating jelly is sterile until the packaging seal is broken 4-oz tube
Catalog No. 50-304-4735 Supplier Med Vet International Supplier No. 1079280EA
Active ingredients include water, propylene glycol and hydroxypropyl methylcellulose HR lubricating jelly is chlorhexidine free Bacteriostatic HR lubricating jelly is sterile until the packaging seal is broken 4-oz tube