Learn More
Med Vet International McKesson Lubricating Jelly, 3-gram Packet, 16-8942, 144/Box
Catalog No. 503042925
Encompass_Preferred
Lubricating Jelly McKesson 3 Gram Individual Packet Sterile Nonstaining and water soluble for quick, easy cleanup Clear, fragrance-free, and stable, nongreasy formula enhances comfort Formulated with improved viscosity to spread easily and evenly Widely used in hospitals and physicians' offices during many routine medical procedures such as catheter insertions and speculum insertions Each packet contains 3 grams of lubricant
Catalog No. 50-304-2925 Supplier Med Vet International Supplier No. 1066698BX
Lubricating Jelly McKesson 3 Gram Individual Packet Sterile Nonstaining and water soluble for quick, easy cleanup Clear, fragrance-free, and stable, nongreasy formula enhances comfort Formulated with improved viscosity to spread easily and evenly Widely used in hospitals and physicians' offices during many routine medical procedures such as catheter insertions and speculum insertions Each packet contains 3 grams of lubricant