Learn More
Med Vet International Lubricating Jelly Aquagel 5 oz. Tube NonSterile, 57-05, Each
Catalog No. 503045495
Encompass_Preferred
Clear, water-soluble, non-irritating lubricant Compatible with natural and synthetic substance Convenient flip cap Hypoallergenic, bacteriostatic 5-ounce tube
Catalog No. 50-304-5495 Supplier Med Vet International Supplier No. 477267EA
Clear, water-soluble, non-irritating lubricant Compatible with natural and synthetic substance Convenient flip cap Hypoallergenic, bacteriostatic 5-ounce tube