Learn More
Med Vet International Lubricating Jelly - Carbomer free Surgilube 4.25 oz. Tube Sterile, 281020537, Each
Catalog No. 503044191
Encompass_Preferred
Surgilube surgical lubricant is non-staining and water-soluble It won't irritate the skin, inner tissue or mucous membranes Surgilube lubricating jelly contains natural gums for improved effectiveness It's suitable for use on instruments or genitals to allow for easy insertion into the body Tip snaps off for single-handed use
Catalog No. 50-304-4191 Supplier Med Vet International Supplier No. 1017415EA
 Restricted Item
Add to Cart
Add to Cart
Surgilube surgical lubricant is non-staining and water-soluble It won't irritate the skin, inner tissue or mucous membranes Surgilube lubricating jelly contains natural gums for improved effectiveness It's suitable for use on instruments or genitals to allow for easy insertion into the body Tip snaps off for single-handed use