Learn More
Med Vet International Miltex Spray Lube, 8oz, Premixed, Non-Silicone, Anti-CorrosIVe Formula, 12/Case
Catalog No. 502829839
Encompass_Preferred
The Miltex 3-700 spray lube is a pre-mixed, non-silicone and anti-corrosive formula in a ready to use 8 ounce pump spray bottle. Steam permeable and water soluble. Helps prevent spotting, staining, and rusting. Lubricates all metal on metal moving parts on surgical instruments.
Catalog No. 50-282-9839 Supplier Med Vet International Supplier No. MIL3700
The Miltex 3-700 spray lube is a pre-mixed, non-silicone and anti-corrosive formula in a ready to use 8 ounce pump spray bottle. Steam permeable and water soluble. Helps prevent spotting, staining, and rusting. Lubricates all metal on metal moving parts on surgical instruments.