Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lubeluzole dihydrochloride | 144665-09-8 | 99.0% | 506.44 | C22H27Cl2F2N3O2S | 1 MG
SDP

Catalog No. 5000243538
Encompass_Preferred

Lubeluzole dihydrochloride is the dihydrochloride salt of lubeluzole, a neuroprotective research reagent that modulates neuronal voltage-gated sodium and calcium channels. It is provided as a high-purity solid intended for in vitro and preclinical pharmacology studies.

  • 99.0% HPLC purity.
  • White to off-white solid appearance.
  • Molecular weight 506.44 g/mol.
  • Molecular formula C22H27Cl2F2N3O2S.
  • Storage: -20°C (sealed); in solution store at -80°C for long-term.
  • Suitable for neuroscience and pharmacology research applications.

Catalog No. 50-002-43538 Supplier Medchemexpress LLC Supplier No. HY105084A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lubeluzole dihydrochloride is the dihydrochloride salt of lubeluzole, a neuroprotective research reagent that modulates neuronal voltage-gated sodium and calcium channels. It is provided as a high-purity solid intended for in vitro and preclinical pharmacology studies.

  • 99.0% HPLC purity.
  • White to off-white solid appearance.
  • Molecular weight 506.44 g/mol.
  • Molecular formula C22H27Cl2F2N3O2S.
  • Storage: -20°C (sealed); in solution store at -80°C for long-term.
  • Suitable for neuroscience and pharmacology research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.