Learn More
Selleck Chemical LLC Lubiprostone S1675-2mg
SDP
Catalog No. 503108467
Encompass_Preferred
Lubiprostone (RU 0211 SPI-0211) is an activator of ClC-2 chloride channels used in the management of idiopathic chronic constipation
Catalog No. 50-310-8467 Supplier Selleck Chemical LLC Supplier No. S16752MG
Lubiprostone (RU 0211 SPI-0211) is an activator of ClC-2 chloride channels used in the management of idiopathic chronic constipation