Learn More
Kano Lab Mfg LUBRCNT AEROKROIL 13OZ 12/CS
SDP
Catalog No. NC9447485
Encompass_Preferred
AeroKroil 13 oz spray,12/cs,It is an industry proven penetrating oil,Quickly loosens rusted nuts and bolts - frees frozen shafts,pulleys,guns,etc,Penetrates to 1millionth inch spaces,dissolves rust,lubricates,cleans and prevents rust
Catalog No. NC9447485 Supplier Kano Lab Mfg Supplier No. KS132C
AeroKroil 13 oz spray,12/cs,It is an industry proven penetrating oil,Quickly loosens rusted nuts and bolts - frees frozen shafts,pulleys,guns,etc,Penetrates to 1millionth inch spaces,dissolves rust,lubricates,cleans and prevents rust