Learn More
Med Vet International Lubricating Jelly HR One Shot 3 Gram Individual Packet Sterile, 208, 30/Box
Catalog No. 503043611
Encompass_Preferred
HR lubricating jelly is chlorhexidine free Bacteriostatic HR lubricating jelly is sterile until the packaging seal is broken 3-gram packets of lubricant ensure you'll have fresh lubricant each time you need it
Catalog No. 50-304-3611 Supplier Med Vet International Supplier No. 1108580BX
HR lubricating jelly is chlorhexidine free Bacteriostatic HR lubricating jelly is sterile until the packaging seal is broken 3-gram packets of lubricant ensure you'll have fresh lubricant each time you need it