Learn More
Med Vet International Lubricating Jelly, E-Z, 2.7gram, Individual Packet, Sterile, MDS032273Z, 144/Box
Catalog No. 503044669
Encompass_Preferred
Sterile, greaseless and water soluble jelly is ideal for lubrication of gloves and instruments Easily spread over rubber, synthetic or metal surfaces, the jelly forms a translucent film that adheres well, yet may be washed away easily
Catalog No. 50-304-4669 Supplier Med Vet International Supplier No. 465515BX
Sterile, greaseless and water soluble jelly is ideal for lubrication of gloves and instruments Easily spread over rubber, synthetic or metal surfaces, the jelly forms a translucent film that adheres well, yet may be washed away easily