Learn More
Med Vet International Lubricating Jelly HR One Shot 3 Gram Individual Packet Sterile, 207, 144/Box
Catalog No. 503045238
Encompass_Preferred
Ingredients include water, propylene glycol and hydroxypropyl methylcellulose Sterile Water soluble Greaseless Unscented Bacteriostatic Box contains a gross of 3-gram packets
Catalog No. 50-304-5238 Supplier Med Vet International Supplier No. 869212BX
Ingredients include water, propylene glycol and hydroxypropyl methylcellulose Sterile Water soluble Greaseless Unscented Bacteriostatic Box contains a gross of 3-gram packets