Learn More
Med Vet International Lubricating Jelly - Carbomer free Surgilube 5 Gram Individual Packet Sterile, 281020545, 144/Box
Catalog No. 503043491
Encompass_Preferred
The Surgilube lubricating jelly has premium viscosity that maintains the integrity of the lube It's water-soluble and doesn't stain fabric The lubricant is pH neutral to soothe skin It's biostatic and will not cause infections It's paraben- and dye-free, ideal for sensitive skin Surgilube is a universal lubricant that can be used topically and internally Lubricating jelly contains no carbomers
Catalog No. 50-304-3491 Supplier Med Vet International Supplier No. 1050804BX
The Surgilube lubricating jelly has premium viscosity that maintains the integrity of the lube It's water-soluble and doesn't stain fabric The lubricant is pH neutral to soothe skin It's biostatic and will not cause infections It's paraben- and dye-free, ideal for sensitive skin Surgilube is a universal lubricant that can be used topically and internally Lubricating jelly contains no carbomers