Learn More
Med Vet International Lubricating Jelly, E-Z, 2.7gram, Individual Packet, Sterile, MDS032273Z, Each
Catalog No. 503044672
Encompass_Preferred
Sterile, greaseless and water soluble jelly is ideal for lubrication of gloves and instruments Easily spread over rubber, synthetic or metal surfaces, the jelly forms a translucent film that adheres well, yet may be washed away easily
Catalog No. 50-304-4672 Supplier Med Vet International Supplier No. 465515EA
Sterile, greaseless and water soluble jelly is ideal for lubrication of gloves and instruments Easily spread over rubber, synthetic or metal surfaces, the jelly forms a translucent film that adheres well, yet may be washed away easily