Learn More
Med Vet International Lubricating Jelly HR One Shot 3 Gram Individual Packet Sterile, 207, Each
Catalog No. 503045240
Encompass_Preferred
Ingredients include water, propylene glycol and hydroxypropyl methylcellulose Sterile Water soluble Greaseless Unscented Bacteriostatic Box contains a gross of 3-gram packets
Catalog No. 50-304-5240 Supplier Med Vet International Supplier No. 869212EA
Ingredients include water, propylene glycol and hydroxypropyl methylcellulose Sterile Water soluble Greaseless Unscented Bacteriostatic Box contains a gross of 3-gram packets