Learn More
Waters Corp Waters Lubricating Oil, 150 cc, HPLC System
Catalog No. 50793532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The oil lubricates the gears of the pump to ensure a smooth flow of solvent through the pump and then to the rest of the system.
  • Designed to lubricate the gears of a pump.
  • Ensures a smooth flow of solvent through the pump and to the rest of the system.
  • Suitable for HPLC systems.
  • Volume: 150 mL.
  • One unit per package.
  • Replacement as required.
Catalog No. 50-793-532 Supplier Waters Corp Supplier No. WAT025661
The oil lubricates the gears of the pump to ensure a smooth flow of solvent through the pump and then to the rest of the system.
  • Designed to lubricate the gears of a pump.
  • Ensures a smooth flow of solvent through the pump and to the rest of the system.
  • Suitable for HPLC systems.
  • Volume: 150 mL.
  • One unit per package.
  • Replacement as required.