Learn More
Waters Corp Waters Corporation Lubrication Kit, Spare Parts
Catalog No. 50793865 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Lubrication Kit from Waters Corporation is an essential component for the maintenance and servicing of Waters equipment. This kit is specifically categorized as a spare parts kit, highlighting its role in ensuring the longevity and optimal performance of compatible instrumentation.
  • Categorized as a Spare Parts Kit
  • Designed for equipment maintenance
  • Used for lubricating the lead screw in compatible Sample Managers
Catalog No. 50-793-865 Supplier Waters Corp Supplier No. WAT272065
The Lubrication Kit from Waters Corporation is an essential component for the maintenance and servicing of Waters equipment. This kit is specifically categorized as a spare parts kit, highlighting its role in ensuring the longevity and optimal performance of compatible instrumentation.
  • Categorized as a Spare Parts Kit
  • Designed for equipment maintenance
  • Used for lubricating the lead screw in compatible Sample Managers