Learn More
MSC Lubriplate No. 105 Grease, NLGI Grade 0, 10 oz
Catalog No. 5000802977
Encompass
This water-resistant lubricant is designed for applications such as plain bearings, cams, and slides, operating up to 140°F. It is also suitable for coating engine parts during assembly and serves as an effective rust preventive grease.
  • Protects freshly machined engine parts
  • Contains zinc-oxide anti-wear additive
  • Prevents dry start, scuffing, scoring, and friction
  • Suitable for motor assembly and rust prevention
  • NLGI grade 0
  • Petroleum-based
  • Operating temperature range -25°F to 140°F
  • White color
Catalog No. 50-008-02977 Supplier MSC Supplier No. 05489612
This water-resistant lubricant is designed for applications such as plain bearings, cams, and slides, operating up to 140°F. It is also suitable for coating engine parts during assembly and serves as an effective rust preventive grease.
  • Protects freshly machined engine parts
  • Contains zinc-oxide anti-wear additive
  • Prevents dry start, scuffing, scoring, and friction
  • Suitable for motor assembly and rust prevention
  • NLGI grade 0
  • Petroleum-based
  • Operating temperature range -25°F to 140°F
  • White color