Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lubrol, MP Biomedicals

Catalog No. ICN19529983 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19529983 250 g
ICN19529980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19529983 Supplier MP Biomedicals, Inc Supplier No. 0219529983
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8047-15-2
Molecular Formula HO(CH2CH2O)n C16H33 n = ∽23
Synonym Polyethylene glycolmonocetyl ether

Specifications

Color White
Physical Form Waxy Solid
Quantity 250 g
Chemical Name or Material Lubrol, Molecular Biology
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.