Learn More
Medchemexpress LLC Luc Yellow CH dilithium | 67769-47-5 | 99.7% | 10 MG
SDP
Catalog No. 5000339243
Encompass_Preferred
Luc Yellow CH dilithium is a high-intensity fluorescent probe containing free hydrazyl groups. It can react with fatty aldehydes at room temperature and serves as a biological tracer to monitor neuronal branching, regeneration, gap junction detection and characterization, and selective ablation of cells after aldehyde fixation. Luc Yellow CH displays the maximum excitation/emission of 430 nm/540 nm, respectively.
  • Fluorescent dye
  • Biological tracer to monitor neuronal branching, regeneration, gap junction detection and characterization, and selective ablation of cells after aldehyde fixation
  • Cell imaging/staining
  • Used for Lucifer yellow CH (10 μg/mL; 12 h) staining images
  • Used to evaluate GJ functionality
Catalog No. 50-003-39243 Supplier Medchemexpress LLC Supplier No. HY12869210MG
Luc Yellow CH dilithium is a high-intensity fluorescent probe containing free hydrazyl groups. It can react with fatty aldehydes at room temperature and serves as a biological tracer to monitor neuronal branching, regeneration, gap junction detection and characterization, and selective ablation of cells after aldehyde fixation. Luc Yellow CH displays the maximum excitation/emission of 430 nm/540 nm, respectively.
  • Fluorescent dye
  • Biological tracer to monitor neuronal branching, regeneration, gap junction detection and characterization, and selective ablation of cells after aldehyde fixation
  • Cell imaging/staining
  • Used for Lucifer yellow CH (10 μg/mL; 12 h) staining images
  • Used to evaluate GJ functionality