Learn More
Aviva Systems Biology Corp LUC7L Antibody - N-terminal region (ARP57815_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57815_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LUC7L. It was validated on Western Blot. LUC7L,LUC7-like (S. cerevisiae),Luc7; SR+89; LUC7B1; hLuc7B1; FLJ10231; LUC7-LIKE; LUC7L,putative RNA-binding protein Luc7-like 1,putative RNA-binding protein Luc7-like 1,OTTHUMP00000067166,OTTHUMP00000194916,putative SR protein LUC7B1,sarcoplasmic reticulum protein LUC7B1The immunogen is a synthetic peptide directed towards the N terminal region of human LUC7L. Alternate names: FLJ10231, LUC7-LIKE, LUC7B1, SR+89, Luc7, hLuc7B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-390