Learn More
Aviva Systems Biology Corp LUC7L Antibody - middle region (ARP57816_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57816_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LUC7L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LUC7L. Alternate names: FLJ10231, LUC7-LIKE, LUC7B1, Luc7, SR+89, hLuc7B1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-391