Learn More
Medchemexpress LLC HY-B2098 10mg Medchemexpress, Lucanthone CAS:479-50-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB209810MG
Encompass_Preferred
Medchemexpress, HY-B2098 10mg Lucanthone CAS:479-50-5 Lucanthone is an endonuclease inhibitor of Apurinic endonuclease-1 ( APE-1 ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3060