Learn More
Lonza Walkersville Lucetta 2 Luminometer
Catalog No. NC1804972
Encompass
The Lucetta 2 Luminometer is a single tube luminometer developed for the detection of bioluminescence and chemiluminescence. It can be used as a portable battery-operated instrument as well as with an external power supply and is suitable for detecting all luminescence. Benefits:Small, portable tube luminometerStand-alone instrument without PC requirementsHigh sensitivity and linearitySpecific measurement mode for running MycoAlert Mycoplasma Detection AssaysQuick single read mode for unprocessed luminescence readingsHigh data storage capacity with maximum storage of up to 2,000 single sample readingsApplications:MycoAlert Mycoplasma Detection AssaysATP assays, e.g. ViaLight Cell Proliferation and Cytotoxicity AssayAdenylate Kinase assays, e.g. ToxiLight Non-destructive Cytotoxicity AssayLuciferase reporter gene analysisContent:1 Lucetta 2 Luminometer instrument
Catalog No. NC1804972 Supplier Lonza Walkersville Supplier No. AAL1002
Add to Cart
Add to Cart
Request a Quote
The Lucetta 2 Luminometer is a single tube luminometer developed for the detection of bioluminescence and chemiluminescence. It can be used as a portable battery-operated instrument as well as with an external power supply and is suitable for detecting all luminescence. Benefits:Small, portable tube luminometerStand-alone instrument without PC requirementsHigh sensitivity and linearitySpecific measurement mode for running MycoAlert Mycoplasma Detection AssaysQuick single read mode for unprocessed luminescence readingsHigh data storage capacity with maximum storage of up to 2,000 single sample readingsApplications:MycoAlert Mycoplasma Detection AssaysATP assays, e.g. ViaLight Cell Proliferation and Cytotoxicity AssayAdenylate Kinase assays, e.g. ToxiLight Non-destructive Cytotoxicity AssayLuciferase reporter gene analysisContent:1 Lucetta 2 Luminometer instrument