Learn More
Sigma Aldrich Fine Chemicals Biosciences Lucfer Yellow CH dpotass100MG
Catalog No. 502904009
Encompass
Lucifer yellow CH is a fixable dye with an excitation and emission spectrum of 430 nm and 540 nm respectively.
Catalog No. 50-290-4009 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L0144100MG
Lucifer yellow CH is a fixable dye with an excitation and emission spectrum of 430 nm and 540 nm respectively.