Learn More
Cayman Chemical LucIdenIc AcId A 10mg
SDP
Catalog No. 502992352
Encompass_Preferred
A triterpene with anticancer activity; cytotoxic to HepG2, HepG2/2.15, and P388 cancer cells (IC50s = 0.164, 0.205, and 17 nM, respectively); inhibits PMA-induced increases in MMP-2 and MMP-9 activity and Matrigel™ invasion in HepG2 cells
Catalog No. 50-299-2352 Supplier Cayman Chemical Supplier No. 3037210 MG
A triterpene with anticancer activity; cytotoxic to HepG2, HepG2/2.15, and P388 cancer cells (IC50s = 0.164, 0.205, and 17 nM, respectively); inhibits PMA-induced increases in MMP-2 and MMP-9 activity and Matrigel™ invasion in HepG2 cells