Learn More
Medchemexpress LLC Lucideric acid A | 95311-94-7 | 1 MG

Supplier: Medchemexpress LLC HYN68621MG
Lucideric acid A is a natural compound isolated from Ganoderma lucidum. It inhibits PMA-induced MMP-9 activity and demonstrates an anti-invasive effect on hepatoma cells.
- Isolated from Ganoderma lucidum
- Inhibits MMP-9 activity
- Anti-invasive effect on hepatoma cells
- Solid, white to off-white appearance
- Molecular formula: C27H38O6
- Molecular weight: 458.59
- Soluble in DMSO (50 mg/mL)
- Store at 4°C, protected from light
- Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.