Learn More
Medchemexpress LLC Lucideric acid A | 95311-94-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN68625MG
Encompass_Preferred
Lucideric acid A is a natural compound isolated from Ganoderma lucidum. It inhibits PMA-induced MMP-9 activity and demonstrates an anti-invasive effect on hepatoma cells.
  • Inhibits MMP-9 activity
  • Shows anti-invasive effect on hepatoma cells
  • Exhibits cytotoxicity against human HeLa, HepG2, and KB cells
  • Exhibits cytotoxicity against mouse P388 cells
  • Inhibits EBV-early antigen activation in human Raji cells
Catalog No. 50-004-19792