Learn More
Biotium Lucifer Yellow CH, lithium salt, 67769-47-5, 25mg
Catalog No. NC1565740
Encompass_Preferred
Molecular Formula C13H9N5O9S2.2Li, Molecular Weight 457.254. Lucifer Yellow CH lithium salt (LY CH lithium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable.
Catalog No. NC1565740 Supplier Biotium Supplier No. 80015
Molecular Formula C13H9N5O9S2.2Li, Molecular Weight 457.254. Lucifer Yellow CH lithium salt (LY CH lithium salt) is a widely used polar molecular tracer for studying neuronal morphology. The fluorescent molecule contains a carbohydrazide that allow the molecule to be aldehyde-fixable.