Learn More
Biotium Lucifer Yellow Cadaverine
Catalog No. NC0943193
Encompass_Preferred
Like Lucifer Yellow CH, Lucifer Yellow Cadaverine can be used as a fixable fluorescent tracer. In addition, the dye can be used to conjugate to carboxylic acid groups.Properties: Ex/ Em nm = 428/536 nm; Yellow solid soluble in water; Store at 4°C and protect from light; C17H17K2N3O8S2; MW: 534
Catalog No. NC0943193 Supplier Biotium Supplier No. 80018
Like Lucifer Yellow CH, Lucifer Yellow Cadaverine can be used as a fixable fluorescent tracer. In addition, the dye can be used to conjugate to carboxylic acid groups.Properties: Ex/ Em nm = 428/536 nm; Yellow solid soluble in water; Store at 4°C and protect from light; C17H17K2N3O8S2; MW: 534