Learn More
FUJIFILM Biosciences Lucifer Yellow CH Dilithium Sa
Catalog No. 5000450247
Encompass_Preferred
Lucifer Yellow CH Dilithium Salt 6-Amino-2- hydrazinocarbonylamino -23-dihydro-13-dioxo-1H-benz de isoquinoline-58-disulfonic acid dilithium salt is fluorescence dye Ex/Em 428/536 of neurons with higher solubility compared with the Lucifer Yellow CH 2K It can be used for studies on morphology of intracellular neurons For research use only RUO
Catalog No. 50-004-50247 Supplier FUJIFILM Biosciences Supplier No. 12506281
Lucifer Yellow CH Dilithium Salt 6-Amino-2- hydrazinocarbonylamino -23-dihydro-13-dioxo-1H-benz de isoquinoline-58-disulfonic acid dilithium salt is fluorescence dye Ex/Em 428/536 of neurons with higher solubility compared with the Lucifer Yellow CH 2K It can be used for studies on morphology of intracellular neurons For research use only RUO