Learn More
Medchemexpress LLC Lucifer yellow CH dilithium salt | 67769-47-5 | MFCD00006922 | 98.4% | 457.25 g/mol | C13H9Li2N5O9S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1286925MG
Encompass_Preferred
Lucifer Yellow CH dilithium salt is a water-soluble, high-intensity fluorescent tracer containing a hydrazide functional group. It is used for intracellular injection, neuronal morphology visualization, gap junction coupling studies, and other applications that require a polar, membrane-impermeant fluorescent probe.
  • High fluorescence intensity for sensitive detection.
  • Water-soluble formulation for easy dissolution in aqueous buffers.
  • Excitation ~428-430 nm and emission ~536-540 nm for common fluorescence setups.
  • Hydrazide group enables reaction with aldehydes for covalent labeling.
  • Solid, yellow to orange appearance suitable for handling and storage.
  • High purity (HPLC) for reproducible experimental performance.
Catalog No. 50-002-62293