Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lucifer Yellow CH Dilithium Salt, Sigma-Aldrich

Catalog No. 01259003 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-003 100 mg
01-258-808 25 mg
01-258-860 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-259-003 Supplier MilliporeSigma Supelco Supplier No. L0259100MG
Add to Cart
Add to Cart

A highly fluorescent dye, useful in marking nerve cells.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lucifer Yellow CH Dilithium Salt
CAS 67769-47-5
Molecular Weight (g/mol) 457.25 g/mol
Color Orange
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.