Learn More
TARGETMOL CHEMICALS INC Lucifer Yellow CH dipotassium
SDP
Catalog No. 502876768
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lucifer Yellow CH dipotassium is a water-soluble fluorescent tracer that labels endocytosis vesicles in cells.Lucifer Yellow CH dipotassium excites and emits at a maximum wavelength of 430 nm and 540 nm, respectively. Purity 99.71%
Catalog No. 50-287-6768 Supplier TARGETMOL CHEMICALS INC Supplier No. T7845410MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Lucifer Yellow CH dipotassium is a water-soluble fluorescent tracer that labels endocytosis vesicles in cells.Lucifer Yellow CH dipotassium excites and emits at a maximum wavelength of 430 nm and 540 nm, respectively. Purity 99.71%