Learn More
Medchemexpress LLC Lucifer yellow CH dipotassium salt | 71206-95-6 | MFCD00013445 | 95.0% | 521.57 g/mol | C13H9K2N5O9S2 | 25 MG
SDP
Catalog No. 5000282818
Encompass_Preferred
Lucifer Yellow CH dipotassium salt is a water-soluble fluorescent tracer dye used for neuronal morphology studies, gap junction detection, and selective cell labeling. Supplied as a stable powder, it is suitable for microscopy and electrophysiology applications where strong, reliable fluorescence is required. The material's solubility and reactive functional groups make it adaptable for fixation and conjugation workflows.
  • Water-soluble fluorescent tracer suitable for neuronal tracing.
  • High fluorescence intensity for clear imaging.
  • Reactive hydrazide groups enable conjugation with aldehydes.
  • Supplied as a stable powder for easy storage and handling.
  • Available in small research-sized quantities (e.g., 25 mg).
Catalog No. 50-002-82818 Supplier Medchemexpress LLC Supplier No. HYW12771525MG
Lucifer Yellow CH dipotassium salt is a water-soluble fluorescent tracer dye used for neuronal morphology studies, gap junction detection, and selective cell labeling. Supplied as a stable powder, it is suitable for microscopy and electrophysiology applications where strong, reliable fluorescence is required. The material's solubility and reactive functional groups make it adaptable for fixation and conjugation workflows.
  • Water-soluble fluorescent tracer suitable for neuronal tracing.
  • High fluorescence intensity for clear imaging.
  • Reactive hydrazide groups enable conjugation with aldehydes.
  • Supplied as a stable powder for easy storage and handling.
  • Available in small research-sized quantities (e.g., 25 mg).